ADVH14DBB315 AChemBlock

1-(2-Methylphenyl)ethylamine

  • MDL number MFCD05215232
  • Cas Number : 42142-17-6
  • Molecular Weight : 135.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-methylphenyl-ethylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38147
Availability details for
Quantity
  • Availability
  • Add to Cart