ADVH14DBB31A AChemBlock

1-(3-Fluorophenyl)ethanamine

  • MDL number MFCD04038308
  • Cas Number : 74788-45-7
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-fluorophenyl-ethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36296
Availability details for
Quantity
  • Availability
  • Add to Cart