ADVH14DBB339 AChemBlock

4'-(Trifluoromethyl)-biphenyl-4-amine

  • MDL number MFCD03424687
  • Cas Number : 57688-34-3
  • Molecular Weight : 237.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-trifluoromethyl-biphenyl-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37289
Availability details for
Quantity
  • Availability
  • Add to Cart