ADVH14DBB370 AChemBlock

N-Cyclopropyl-4-(trifluoromethyl)-benzylamine

  • MDL number MFCD09043527
  • Cas Number : 643007-99-2
  • Molecular Weight : 215.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-cyclopropyl-4-trifluoromethyl-benzylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36867
Availability details for
Quantity
  • Availability
  • Add to Cart