ADVH14DBB3C0 AChemBlock

tert-Butyl [2-(3-aminophenoxy)ethyl]carbamate

  • MDL number MFCD11922407
  • Cas Number : 246240-10-8
  • Molecular Weight : 252.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-3-aminophenoxy-ethyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39402
Availability details for
Quantity
  • Availability
  • Add to Cart