ADVH14DBB3CE AChemBlock

4-(4-((Dimethylamino)methyl)piperidin-1-yl)aniline

  • MDL number MFCD16251975
  • Cas Number : 1224932-55-1
  • Molecular Weight : 233.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-dimethylamino-methyl-piperidin-1-yl-aniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42171
Availability details for
Quantity
  • Availability
  • Add to Cart