ADVH14DBB3D4 AChemBlock

1-(2-Morpholin-4-yl-ethyl)-1H-pyrazol-4-ylamine

  • MDL number MFCD09965621
  • Cas Number : 1152961-27-7
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-morpholin-4-yl-ethyl-1h-pyrazol-4-ylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42522
Availability details for
Quantity
  • Availability
  • Add to Cart