ADVH14DBB441 AChemBlock

3-(Benzyloxy)-N-methylpropan-1-amine

  • MDL number MFCD16662989
  • Cas Number : 188878-41-3
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-benzyloxy-n-methylpropan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40223
Availability details for
Quantity
  • Availability
  • Add to Cart