ADVH14DBB47A AChemBlock

2-(2-Fluoro-4-methoxyphenyl)ethanamine hydrochloride

  • MDL number MFCD28978585
  • Cas Number : 402832-76-2
  • Molecular Weight : 205.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-fluoro-4-methoxyphenyl-ethanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38246
Availability details for
Quantity
  • Availability
  • Add to Cart