ADVH14DBB47D AChemBlock

(3-Phenoxy)propylamine

  • MDL number MFCD04038049
  • Cas Number : 7617-76-7
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-phenoxy-propylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36207
Availability details for
Quantity
  • Availability
  • Add to Cart