ADVH14DBB4A4 AChemBlock

2-(1H-Imidazol-1-ylmethyl)-morpholine trihydrochloride

  • MDL number MFCD31560286
  • Cas Number : 1341869-57-5
  • Molecular Weight : 276.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-1h-imidazol-1-ylmethyl-morpholine-trihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41601
Availability details for
Quantity
  • Availability
  • Add to Cart