ADVH14DBB4D0 AChemBlock

3-(3-Methoxyphenyl)pyrrolidine hydrochloride

  • MDL number MFCD03701257
  • Cas Number : 38175-35-8
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-3-methoxyphenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38409
Availability details for
Quantity
  • Availability
  • Add to Cart