ADVH14DBB4D5 AChemBlock

3-(4-Bromophenyl)pyrrolidine hydrochloride

  • MDL number MFCD09997901
  • Cas Number : 1187931-39-0
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-bromophenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42400
Availability details for
Quantity
  • Availability
  • Add to Cart