ADVH14DBB507 AChemBlock

6-(3-Methoxyphenyl)-3-pyridinecarboxaldehyde

  • MDL number MFCD04115432
  • Cas Number : 898405-27-1
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-3-methoxyphenyl-3-pyridinecarboxaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35299
Availability details for
Quantity
  • Availability
  • Add to Cart