ADVH14DBB529 AChemBlock

4'-Chloro-[1,1'-biphenyl]-3-ol

  • MDL number MFCD00871469
  • Cas Number : 28023-90-7
  • Molecular Weight : 204.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-1-1-biphenyl-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39091
Availability details for
Quantity
  • Availability
  • Add to Cart