ADVH14DBB52C AChemBlock

2-Benzyl-3-phenylpropanoic acid

  • MDL number MFCD00796480
  • Cas Number : 618-68-8
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-benzyl-3-phenylpropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37040
Availability details for
Quantity
  • Availability
  • Add to Cart