ADVH14DBB576 AChemBlock

1-(3-Bromopropyl)-3,5-difluorobenzene

  • MDL number MFCD09744393
  • Cas Number : 872414-38-5
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-bromopropyl-3-5-difluorobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35595
Availability details for
Quantity
  • Availability
  • Add to Cart