ADVH14DBB593 AChemBlock

tert-Butyl cyclopropanecarboxylate

  • MDL number MFCD09836696
  • Cas Number : 87661-20-9
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-cyclopropanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35556
Availability details for
Quantity
  • Availability
  • Add to Cart