ADVH14DBB5C8 AChemBlock

3-Methoxy-phenyl propargyl ether

  • MDL number MFCD14631228
  • Cas Number : 41580-72-7
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methoxy-phenyl-propargyl-ether.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38172
Availability details for
Quantity
  • Availability
  • Add to Cart