ADVH14DBB5FA AChemBlock

3-Amino-1-(1-piperidinyl)-1-propanone hydrochloride

  • MDL number MFCD08461913
  • Cas Number : 161862-09-5
  • Molecular Weight : 192.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-1-1-piperidinyl-1-propanone-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40818
Availability details for
Quantity
  • Availability
  • Add to Cart