ADVH14DBB612 AChemBlock

3-(2-Methylphenoxy)piperidine hydrochloride

  • MDL number MFCD21606599
  • Cas Number : 902837-28-9
  • Molecular Weight : 227.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-methylphenoxy-piperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35265
Availability details for
Quantity
  • Availability
  • Add to Cart