ADVH14DBB626 AChemBlock

1-Boc-4-hydroxy-4-methyl-piperidine

  • MDL number MFCD04972471
  • Cas Number : 406235-30-1
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-4-hydroxy-4-methyl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38205
Availability details for
Quantity
  • Availability
  • Add to Cart