ADVH14DBB667 AChemBlock

Methyl 2,4-difluoro-b-oxo-benzenebutanate

  • MDL number MFCD08457114
  • Cas Number : 942475-04-9
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-4-difluoro-b-oxo-benzenebutanate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34938
Availability details for
Quantity
  • Availability
  • Add to Cart