ADVH14DBB686 AChemBlock

3-Oxo-5-phenylpentanoic acid methyl ester

  • MDL number MFCD11849521
  • Cas Number : 30414-58-5
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-oxo-5-phenylpentanoic-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38937
Availability details for
Quantity
  • Availability
  • Add to Cart