ADVH14DBB691 AChemBlock

Ethyl 3-(2,3-dichlorophenyl)-3-oxopropanoate

  • MDL number MFCD09756184
  • Cas Number : 672323-07-8
  • Molecular Weight : 261.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-2-3-dichlorophenyl-3-oxopropanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36703
Availability details for
Quantity
  • Availability
  • Add to Cart