ADVH14DBB6AB AChemBlock

Ethyl 2-methoxy-a,g-dioxo-benzenebutanoate

  • MDL number MFCD08700415
  • Cas Number : 60943-39-7
  • Molecular Weight : 250.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-methoxy-a-g-dioxo-benzenebutanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37105
Availability details for
Quantity
  • Availability
  • Add to Cart