ADVH14DBB6C0 AChemBlock

1-[4-(Trifluoromethyl)phenyl]-1,3-butanedione

  • MDL number MFCD07772941
  • Cas Number : 258346-69-9
  • Molecular Weight : 230.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-trifluoromethyl-phenyl-1-3-butanedione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39295
Availability details for
Quantity
  • Availability
  • Add to Cart