ADVH14DBB73E AChemBlock

(S)-2-Amino-5-hexenoic acid

  • MDL number MFCD08272861
  • Cas Number : 90989-12-1
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-5-hexenoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35206
Availability details for
Quantity
  • Availability
  • Add to Cart