ADVH14DBB74A AChemBlock

N-Boc-D-phenylalanine N'-methoxy-N'-methylamide

  • MDL number MFCD18252762
  • Cas Number : 115186-33-9
  • Molecular Weight : 308.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-d-phenylalanine-n-methoxy-n-methylamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42526
Availability details for
Quantity
  • Availability
  • Add to Cart