ADVH14DBB754 AChemBlock

Ethyl 3-(aminomethyl)benzoate hydrochloride

  • MDL number MFCD16660252
  • Cas Number : 91843-34-4
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-aminomethyl-benzoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35086
Availability details for
Quantity
  • Availability
  • Add to Cart