ADVH14DBB766 AChemBlock

Fmoc-4-(aminomethyl)-oxan-4-carboxylic acid

  • MDL number MFCD09264209
  • Cas Number : 946716-29-6
  • Molecular Weight : 381.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-4-aminomethyl-oxan-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34897
Availability details for
Quantity
  • Availability
  • Add to Cart