ADVH14DBB770 AChemBlock

3-Amino-L-alanine methyl ester dihydrochloride

  • MDL number MFCD09998031
  • Cas Number : 147857-43-0
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-l-alanine-methyl-ester-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41137
Availability details for
Quantity
  • Availability
  • Add to Cart