ADVH14DBB782 AChemBlock

3-(Boc-amino)-DL-alanine

  • MDL number MFCD21090341
  • Cas Number : 96895-04-4
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-boc-amino-dl-alanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34784
Availability details for
Quantity
  • Availability
  • Add to Cart