ADVH14DBB7A3 AChemBlock

Methyl 2-acetamido-2-(dimethoxyphosphoryl)acetate

  • MDL number MFCD08443722
  • Cas Number : 89524-99-2
  • Molecular Weight : 239.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-acetamido-2-dimethoxyphosphoryl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35314
Availability details for
Quantity
  • Availability
  • Add to Cart