ADVH14DBB7F1 AChemBlock

1-Phenylcyclobutanamine

  • MDL number MFCD08444192
  • Cas Number : 17380-77-7
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-phenylcyclobutanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40536
Availability details for
Quantity
  • Availability
  • Add to Cart