ADVH14DBB8C5 AChemBlock

1-(3,4-Difluorobenzyl)piperazine dihydrochloride

  • MDL number MFCD30729921
  • Cas Number : 203860-01-9
  • Molecular Weight : 285.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-4-difluorobenzyl-piperazine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39950
Availability details for
Quantity
  • Availability
  • Add to Cart