ADVH14DBB8F3 AChemBlock

3-Piperazin-1-yl-benzoic acid

  • MDL number MFCD03444416
  • Cas Number : 446831-28-3
  • Molecular Weight : 206.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-piperazin-1-yl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38039
Availability details for
Quantity
  • Availability
  • Add to Cart