ADVH14DBB917 AChemBlock

(S)-4-Benzyl-2-hydroxymethylpiperazine

  • MDL number MFCD11113047
  • Cas Number : 149715-45-7
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-benzyl-2-hydroxymethylpiperazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41091
Availability details for
Quantity
  • Availability
  • Add to Cart