ADVH14DBB918 AChemBlock

(R)-4-Benzyl-2-hydroxymethylpiperazine

  • MDL number MFCD11113046
  • Cas Number : 149715-46-8
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-4-benzyl-2-hydroxymethylpiperazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41090
Availability details for
Quantity
  • Availability
  • Add to Cart