ADVH14DBB93C AChemBlock

1,4-Dihydro-3(2H)-isoquinolinone

  • MDL number MFCD03460354
  • Cas Number : 24331-94-0
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-dihydro-3-2h-isoquinolinone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39429
Availability details for
Quantity
  • Availability
  • Add to Cart