ADVH14DBB956 AChemBlock

3-(Methylamino)sulfolane hydrochloride

  • MDL number MFCD00456571
  • Cas Number : 53287-53-9
  • Molecular Weight : 185.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methylamino-sulfolane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37594
Availability details for
Quantity
  • Availability
  • Add to Cart