ADVH14DBB9C5 AChemBlock

3-Amino-6-methoxy-2-methylpyridine

  • MDL number MFCD03095090
  • Cas Number : 52090-56-9
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-6-methoxy-2-methylpyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37660
Availability details for
Quantity
  • Availability
  • Add to Cart