ADVH14DBB9D9 AChemBlock

1-Cyclopropyl-4,4,4-trifluoro-1,3-butanedione

  • MDL number MFCD03419771
  • Cas Number : 30923-69-4
  • Molecular Weight : 180.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-cyclopropyl-4-4-4-trifluoro-1-3-butanedione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38909
Availability details for
Quantity
  • Availability
  • Add to Cart