ADVH14DBBA01 AChemBlock

b-Oxo-1-naphthalenepropanoic acid ethyl ester

  • MDL number MFCD03424836
  • Cas Number : 62071-76-5
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/b-oxo-1-naphthalenepropanoic-acid-ethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37027
Availability details for
Quantity
  • Availability
  • Add to Cart