ADVH14DBBA16 AChemBlock

1-(4-Fluorophenyl)-1,3-butanedione

  • MDL number MFCD00463139
  • Cas Number : 29681-98-9
  • Molecular Weight : 180.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-fluorophenyl-1-3-butanedione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39000
Availability details for
Quantity
  • Availability
  • Add to Cart