ADVH14DBBA3F AChemBlock

3-Iodo-benzenepropanoic acid

  • MDL number MFCD03425691
  • Cas Number : 68034-75-3
  • Molecular Weight : 276.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-benzenepropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36632
Availability details for
Quantity
  • Availability
  • Add to Cart