ADVH14DBBA45 AChemBlock

1-(3-Trifluoromethylphenyl)-1-cyanocyclopropane

  • MDL number MFCD07374416
  • Cas Number : 124305-68-6
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-trifluoromethylphenyl-1-cyanocyclopropane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42086
Availability details for
Quantity
  • Availability
  • Add to Cart