ADVH14DBBA56 AChemBlock

3-Thiophenepropanoic acid

  • MDL number MFCD06655385
  • Cas Number : 16378-06-6
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-thiophenepropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40765
Availability details for
Quantity
  • Availability
  • Add to Cart