ADVH14DBBA5B AChemBlock

(R)-a-Hydroxy-cyclohexanepropanoic acid

  • MDL number MFCD17171007
  • Cas Number : 156469-00-0
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-a-hydroxy-cyclohexanepropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40940
Availability details for
Quantity
  • Availability
  • Add to Cart