ADVH14DBBA93 AChemBlock

2-(2-Aminophenoxy)acetonitrile

  • MDL number MFCD09812626
  • Cas Number : 31507-29-6
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-aminophenoxy-acetonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38881
Availability details for
Quantity
  • Availability
  • Add to Cart